Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012181091.2 | XP_012036481.1 | 3 - 4 | 803 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |