Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976542.1 | XP_027832343.1 | 2 - 3 | 751 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |