Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957471.1 | XP_027813272.1 | 7 - 8 | 883 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |