Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012818.4 | XP_004012867.2 | 3 - 4 | 773 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |