Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012179896.3 | XP_012035286.1 | 1 - 2 | 87 | 108 | |
XM_012179898.3 | XP_012035288.2 | 1 - 2 | 119 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |