Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022221.4 | XP_004022270.1 | 2 - 3 | 447 | 102 | |
XM_012106989.3 | XP_011962379.1 | 2 - 3 | 460 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |