Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012095281.2 | XP_011950671.2 | 5 - 6 | 619 | 103 | |
XM_012095282.3 | XP_011950672.2 | 5 - 6 | 583 | 103 | ||
XM_012095283.2 | XP_011950673.2 | 3 - 4 | 245 | 103 | ||
XM_012095284.2 | XP_011950674.2 | 2 - 3 | 173 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |