Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015074.3 | XP_004015123.2 | 2 - 3 | 668 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |