Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015097.4 | XP_004015146.2 | 4 - 5 | 525 | 112 | |
XM_012190032.3 | XP_012045422.2 | 4 - 5 | 464 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |