Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTEX1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096392.3 | XP_011951782.2 | 2 - 3 | 458 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |