Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097742.2 | XP_014953228.2 | 6 - 7 | 921 | 107 | |
XM_027972914.1 | XP_027828715.1 | 7 - 8 | 1004 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |