Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980182.1 | XP_027835983.1 | 3 - 4 | 1372 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |