Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975349.1 | XP_027831150.1 | 5 - 6 | 1119 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |