Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957386.1 | XP_027813187.1 | 3 - 4 | 1182 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |