Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966026.1 | XP_027821827.1 | 6 - 7 | 1186 | 63 | |
XM_027966027.1 | XP_027821828.1 | 7 - 8 | 921 | 63 | ||
XM_027966028.1 | XP_027821829.1 | 7 - 8 | 920 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |