Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002705.4 | XP_004002754.1 | 1 - 2 | 683 | 68 | |
XM_012185028.3 | XP_012040418.1 | 1 - 2 | 684 | 68 | ||
XM_012185031.3 | XP_012040421.1 | 1 - 2 | 684 | 68 | ||
XM_012185041.3 | XP_012040431.1 | 1 - 2 | 683 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |