Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968305.1 | XP_027824106.1 | 2 - 3 | 241 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |