Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015325.4 | XP_004015374.2 | 2 - 3 | 387 | 89 | |
XM_027978516.1 | XP_027834317.1 | 2 - 3 | 627 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |