Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100804.3 | XP_011956194.1 | 7 - 8 | 936 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |