Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107263.3 | XP_011962653.2 | 4 - 5 | 909 | 96 | |
XM_012107264.3 | XP_011962654.2 | 4 - 5 | 920 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |