Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974669.1 | XP_027830470.1 | 11 - 12 | 3980 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |