Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015267.4 | XP_004015316.2 | 8 - 9 | 1345 | 72 | |
GenBank mRNA | HQ263377.1 | - | 8 - 9 | 1330 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |