Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973706.1 | XP_027829507.1 | 3 - 4 | 1028 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |