Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100263.2 | XP_014955749.2 | 7 | 1868 | 89 | |
XM_027978561.1 | XP_027834362.1 | 7 | 1980 | 89 | ||
XM_027978562.1 | XP_027834363.1 | 7 | 2002 | 89 | ||
XM_027978563.1 | XP_027834364.1 | 7 | 1890 | 89 | ||
XM_027978564.1 | XP_027834365.1 | 7 | 1965 | 89 | ||
XM_027978565.1 | XP_027834366.1 | 7 | 1943 | 89 | ||
XM_027978566.1 | XP_027834367.1 | 6 | 1837 | 89 | ||
XM_027978567.1 | XP_027834368.1 | 6 | 2028 | 89 | ||
XM_027978568.1 | XP_027834369.1 | 5 | 1557 | 89 | ||
XM_027978569.1 | XP_027834370.1 | 6 | 1646 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |