Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964707.1 | XP_027820508.1 | 4 - 5 | 1988 | 130 | |
XM_027964708.1 | XP_027820509.1 | 4 - 5 | 1988 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |