Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104397.3 | XP_011959787.2 | 2 | 979 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |