Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC4A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082005.1 | NP_001075474.1 | 1 - 2 | 238 | 66 | |
XM_008254828.2 | XP_008253050.1 | 1 - 2 | 836 | 66 | ||
XM_008254829.2 | XP_008253051.1 | 1 - 2 | 837 | 66 | ||
XM_017340967.1 | XP_017196456.1 | 1 - 2 | 837 | 66 | ||
XM_017340968.1 | XP_017196457.1 | 1 - 2 | 838 | 66 | ||
XM_017340969.1 | XP_017196458.1 | 1 - 2 | 837 | 66 | ||
XM_017340970.1 | XP_017196459.1 | 1 - 2 | 838 | 66 | ||
XM_017340971.1 | XP_017196460.1 | 1 - 2 | 837 | 66 | ||
XM_017340972.1 | XP_017196461.1 | 1 - 2 | 838 | 66 | ||
XM_017340973.1 | XP_017196462.1 | 1 - 2 | 840 | 66 | ||
XM_017340975.1 | XP_017196464.1 | 1 - 2 | 840 | 66 | ||
GenBank mRNA | AB038263.1 | - | 1 - 2 | 238 | 66 | |
AB038264.1 | - | 1 - 2 | 238 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |