Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082008.1 | NP_001075477.1 | 1 - 2 | 95 | 91 | |
XM_008260447.2 | XP_008258669.1 | 1 - 2 | 124 | 91 | ||
GenBank mRNA | AB050593.1 | - | 1 - 2 | 95 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |