Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082044.1 | NP_001075513.1 | 36 - 37 | 4472 | 88 | |
XM_008259331.2 | XP_008257553.1 | 37 - 38 | 4486 | 88 | ||
XM_008259332.2 | XP_008257554.1 | 37 - 38 | 4485 | 88 | ||
XM_008259335.2 | XP_008257557.1 | 36 - 37 | 4446 | 88 | ||
XM_008259336.2 | XP_008257558.1 | 36 - 37 | 4447 | 88 | ||
XM_017343242.1 | XP_017198731.1 | 37 - 38 | 4488 | 88 | ||
GenBank mRNA | AF087468.1 | - | 36 - 37 | 4472 | 88 | |
AF417510.1 | - | 34 - 35 | 4199 | 88 | ||
AF417511.1 | - | 34 - 35 | 4199 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |