Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082118.1 | NP_001075587.1 | 5 - 6 | 1720 | 95 | |
GenBank mRNA | AF493547.1 | - | 5 - 6 | 1680 | 95 | |
AF508735.1 | - | 5 - 6 | 1720 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |