Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082119.1 | NP_001075588.1 | 5 - 6 | 491 | 89 | |
XM_008255642.2 | XP_008253864.1 | 4 - 5 | 444 | 89 | ||
GenBank mRNA | AF510726.1 | - | 5 - 6 | 491 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |