Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082136.1 | NP_001075605.1 | 1 - 2 | 251 | 124 | |
GenBank mRNA | AY013262.1 | - | 1 - 2 | 251 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Oryctolagus cuniculus Probe spans exons |