Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082138.1 | NP_001075607.1 | 1 - 2 | 112 | 94 | |
XM_008263951.2 | XP_008262173.1 | 3 - 4 | 268 | 94 | ||
XM_008263952.2 | XP_008262174.1 | 2 - 3 | 223 | 94 | ||
GenBank mRNA | AY050567.1 | - | 1 - 2 | 112 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |