Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082206.1 | NP_001075675.1 | 15 - 16 | 1820 | 87 | |
GenBank mRNA | D45239.1 | - | 15 - 16 | 1820 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Inventoried Probe spans exons |