Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082230.1 | NP_001075699.1 | 2 - 3 | 293 | 67 | |
GenBank mRNA | J03744.1 | - | 2 - 3 | 293 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |