Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082284.1 | NP_001075753.1 | 2 - 3 | 213 | 109 | |
XM_008263973.2 | XP_008262195.2 | 2 - 3 | 257 | 109 | ||
GenBank mRNA | M32029.1 | - | 2 - 3 | 213 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |