Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082303.1 | NP_001075772.1 | 3 - 4 | 977 | 138 | |
GenBank mRNA | M65109.1 | - | 3 - 4 | 977 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |