Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082310.1 | NP_001075779.1 | 5 - 6 | 670 | 62 | |
GenBank mRNA | M84474.1 | - | 5 - 6 | 670 | 62 | |
M84988.1 | - | 5 - 6 | 670 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |