Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082370.1 | NP_001075839.1 | 7 - 8 | 1100 | 92 | |
GenBank mRNA | U57553.1 | - | 7 - 8 | 1066 | 92 | |
X12661.1 | - | 742 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |