Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082405.1 | NP_001075874.1 | 1430 | 99 | ||
GenBank mRNA | X91902.1 | - | 1430 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus |