Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 5 - 6 | 544 | 81 | |
XM_017346648.1 | XP_017202137.1 | 4 - 5 | 511 | 81 | ||
GenBank mRNA | U32507.1 | - | 5 - 6 | 544 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |