Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082710.1 | NP_001076179.1 | 1689 | 59 | ||
GenBank mRNA | AF005355.1 | - | 1689 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus |