Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082750.1 | NP_001076219.1 | 19 - 20 | 3004 | 99 | |
NM_001170980.1 | NP_001164451.1 | 8 - 9 | 870 | 99 | ||
XM_008259342.2 | XP_008257564.1 | 20 - 21 | 3127 | 99 | ||
XM_017343246.1 | XP_017198735.1 | 18 - 19 | 2920 | 99 | ||
GenBank mRNA | U09490.1 | - | 19 - 20 | 3004 | 99 | |
U32587.1 | - | 8 - 9 | 870 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |