Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082787.2 | NP_001076256.1 | 1 - 2 | 173 | 93 | |
XM_008255647.2 | XP_008253869.1 | 2 - 3 | 368 | 93 | ||
GenBank mRNA | AY101395.1 | - | 1 - 2 | 175 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Oryctolagus cuniculus Probe spans exons |