Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164863.1 | NP_001158335.1 | 5 - 6 | 683 | 66 | |
XM_008252716.2 | XP_008250938.1 | 6 - 7 | 1232 | 66 | ||
GenBank mRNA | EU731009.1 | - | 5 - 6 | 683 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |