Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160282.1 | NP_001153754.1 | 2 - 3 | 484 | 83 | |
XM_008264005.2 | XP_008262227.1 | 3 - 4 | 926 | 83 | ||
GenBank mRNA | FJ028656.1 | - | 2 - 3 | 484 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |