Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082199.1 | NP_001075668.1 | 2 - 3 | 307 | 72 | |
GenBank mRNA | D21264.1 | - | 2 - 3 | 307 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |