Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171042.1 | NP_001164513.1 | 3 - 4 | 864 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |