Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLA-DR-ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171118.1 | NP_001164589.1 | 3 - 4 | 677 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |