Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171141.1 | NP_001164612.1 | 1 | 1443 | 86 | |
GenBank mRNA | M99311.1 | - | 1 | 1431 | 86 | |
M99312.1 | - | 1 | 1431 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |